AMBH5806421D Ambeed, Inc.

(1S,4R)-4-Propyl-4'-(4-(trifluoromethoxy)phenyl)-1,1'-bi(cyclohexane)

  • MDL number MFCD20488041
  • Cas Number : 133937-72-1
  • Molecular Weight : 368.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/133937-72-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/133937-72-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart