AMBH58064224 Ambeed, Inc.

tert-Butyl 3-iodo-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate

  • MDL number MFCD28501441
  • Cas Number : 657428-55-2
  • Molecular Weight : 335.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-iodo-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate
Inchi Code 1S/C10H14IN3O2/c1-10(2,3)16-9(15)14-4-6-7(5-14)12-13-8(6)11/h4-5H2,1-3H3,(H,12,13)
Inchi Key YTTAUUCKFOALEH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/657428-55-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/657428-55-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart