AMBH5806422A Ambeed, Inc.

2-Chloro-1-isopropyl-1H-benzo[d]imidazole

  • MDL number MFCD00456409
  • Cas Number : 3705-87-1
  • Molecular Weight : 194.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/3705-87-1.html

Safety Information

MSDS https://www.ambeed.com/products/3705-87-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart