AMBH58064233 Ambeed, Inc.

(3S,4R,5S)-Methyl 3,4,5,6-tetrahydroxy-2-oxohexanoate

  • MDL number MFCD00021987
  • Cas Number : 3031-98-9
  • Molecular Weight : 208.1700

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/3031-98-9.html

Safety Information

signalword Warning
pcodes P305+P351+P338-P280
MSDS https://www.ambeed.com/products/3031-98-9.html
pictograms GHS07
hstatements H319-H317
Availability details for
Quantity
  • Availability
  • Add to Cart