AMBH5806423D Ambeed, Inc.

Bis(2-ethylhexyl)carbamodithioic acid

  • MDL number N/A
  • Cas Number : 77414-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/77414-73-4.html

Safety Information

MSDS https://www.ambeed.com/products/77414-73-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart