AMBH58064245 Ambeed, Inc.

tert-Butyl (2-chloroethyl)(methyl)carbamate

  • MDL number MFCD20624610
  • Cas Number : 220074-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form Liquid or Solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/220074-38-4.html

Safety Information

MSDS https://www.ambeed.com/products/220074-38-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart