AMBH5806424B Ambeed, Inc.

4-(4-Chlorophenyl)dihydro-2H-pyran-2,6(3H)-dione

  • MDL number MFCD00190250
  • Cas Number : 53911-68-5
  • Molecular Weight : 224.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/53911-68-5.html

Safety Information

MSDS https://www.ambeed.com/products/53911-68-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart