AMBH5806424D Ambeed, Inc.

2-Methyl-5-(methylsulfonyl)-1,3,4-oxadiazole

  • MDL number MFCD06496480
  • Cas Number : 205646-91-9
  • Molecular Weight : 162.1700

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/205646-91-9.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/205646-91-9.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart