AMBH58064258 Ambeed, Inc.

4-(Bromomethyl)-3-methylpyridine hydrobromide

  • MDL number MFCD30527969
  • Cas Number : 2201551-69-9
  • Molecular Weight : 266.9611

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2201551-69-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/2201551-69-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart