AMBH5806425D Ambeed, Inc.

4-Bromo-6-methoxypyrimidin-2-amine

  • MDL number MFCD20486653
  • Cas Number : 53557-85-0
  • Molecular Weight : 204.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/53557-85-0.html

Safety Information

MSDS https://www.ambeed.com/products/53557-85-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart