AMBH58064261 Ambeed, Inc.

Methyl 6-amino-5-chloropicolinate

  • MDL number MFCD18250740
  • Cas Number : 1256794-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1256794-05-4.html

Safety Information

MSDS https://www.ambeed.com/products/1256794-05-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart