AMBH58064264 Ambeed, Inc.

Methyl 1-O-(2-formyl-4-nitrophenyl)-2,3,4-tri-O-acetyl-�D-glucopyranuronate

  • MDL number N/A
  • Cas Number : 148579-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/148579-83-3.html

Safety Information

MSDS https://www.ambeed.com/products/148579-83-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart