AMBH58064266 Ambeed, Inc.

4-Methylphenyl tetra-O-acetyl-a-D-galactopyranoside

  • MDL number N/A
  • Cas Number : 52730-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.ambeed.com/products/52730-14-0.html

Safety Information

MSDS https://www.ambeed.com/products/52730-14-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart