AMBH5806426B Ambeed, Inc.

N-(3-Hydroxyphenyl)pivalamide

  • MDL number N/A
  • Cas Number : 75151-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/75151-82-5.html

Safety Information

MSDS https://www.ambeed.com/products/75151-82-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart