AMBH5806426D Ambeed, Inc.

Ethyl 2-(bromotriphenyl-l5-phosphanyl)acetate

  • MDL number N/A
  • Cas Number : 4083-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/4083-85-6.html

Safety Information

MSDS https://www.ambeed.com/products/4083-85-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart