AMBH5806426F Ambeed, Inc.

2,2'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl

  • MDL number MFCD30729880
  • Cas Number : 398128-09-1
  • Molecular Weight : 406.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2'-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl
Inchi Code 1S/C24H32B2O4/c1-21(2)22(3,4)28-25(27-21)19-15-11-9-13-17(19)18-14-10-12-16-20(18)26-29-23(5,6)24(7,8)30-26/h9-16H,1-8H3
Inchi Key ZIKMBZIYSBFMMV-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/398128-09-1.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/398128-09-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart