AMBH5806427F Ambeed, Inc.

KN-92 hydrochloride

  • MDL number MFCD25372041
  • Cas Number : 1431698-47-3
  • Molecular Weight : 493.4458

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1431698-47-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1431698-47-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart