AMBH58064283 Ambeed, Inc.

Cinnamyl formate

  • MDL number MFCD00021048
  • Cas Number : 104-65-4
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/104-65-4.html

Safety Information

signalword Warning
pcodes P261-P280
MSDS https://www.ambeed.com/products/104-65-4.html
pictograms GHS07
hstatements H317
Availability details for
Quantity
  • Availability
  • Add to Cart