AMBH58064288 Ambeed, Inc.

5-Methyl-2-(prop-1-en-2-yl)cyclohexan-1-ol

  • MDL number MFCD00001479
  • Cas Number : 7786-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/7786-67-6.html

Safety Information

MSDS https://www.ambeed.com/products/7786-67-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart