AMBH58064291 Ambeed, Inc.

3-Bromo-1-(4-fluorophenyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD16683349
  • Cas Number : 1251249-98-5
  • Molecular Weight : 285.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1251249-98-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1251249-98-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart