AMBH58064292 Ambeed, Inc.

2-(Prop-2-en-1-yl)benzaldehyde

  • MDL number MFCD18824313
  • Cas Number : 62708-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/62708-42-3.html

Safety Information

MSDS https://www.ambeed.com/products/62708-42-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart