AMBH58064294 Ambeed, Inc.

3-Chloro-5-(chlorosulfonyl)-4-hydroxybenzoic acid

  • MDL number MFCD18839087
  • Cas Number : 1201663-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1201663-81-1.html

Safety Information

MSDS https://www.ambeed.com/products/1201663-81-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart