AMBH58064295 Ambeed, Inc.

(R)-Benzyl 2-amino-3-(3-(methylsulfonyl)phenyl)propanoate hydrochloride

  • MDL number MFCD30570304
  • Cas Number : 2049127-88-8
  • Molecular Weight : 369.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2049127-88-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2049127-88-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart