AMBH58064296 Ambeed, Inc.

3-(Bromomethyl)-4-methylthiophene

  • MDL number MFCD28670238
  • Cas Number : 856937-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/856937-70-7.html

Safety Information

MSDS https://www.ambeed.com/products/856937-70-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart