AMBH58064297 Ambeed, Inc.

(1-(2-Aminoethyl)-1H-pyrazol-4-yl)boronic acid hydrochloride

  • MDL number N/A
  • Molecular Weight : 191.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1-(2-aminoethyl)-1H-pyrazol-4-yl)boronic acid hydrochloride
Inchi Code 1S/C5H10BN3O2.ClH/c7-1-2-9-4-5(3-8-9)6(10)11;/h3-4,10-11H,1-2,7H2;1H
Inchi Key LCLDPWIKRCZPAH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/P001052031.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/P001052031.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart