AMBH5806429F Ambeed, Inc.

2-(6-Chloropyridin-3-yl)ethanol

  • MDL number MFCD15526719
  • Cas Number : 117528-28-6
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(6-chloro-3-pyridinyl)ethanol
Inchi Code 1S/C7H8ClNO/c8-7-2-1-6(3-4-10)5-9-7/h1-2,5,10H,3-4H2
Inchi Key GRVARUXBHWDCML-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/117528-28-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/117528-28-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart