AMBH580642A0 Ambeed, Inc.

Methyl 2-methyl-6-nitro-2H-indazole-3-carboxylate

  • MDL number N/A
  • Cas Number : 1058740-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/1058740-89-8.html

Safety Information

MSDS https://www.ambeed.com/products/1058740-89-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart