AMBH580642A2 Ambeed, Inc.

4-Bromo-1-isopropyl-2-methoxybenzene

  • MDL number MFCD28356548
  • Cas Number : 1369775-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/1369775-86-9.html

Safety Information

MSDS https://www.ambeed.com/products/1369775-86-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart