AMBH580642A3 Ambeed, Inc.

4-(((5-Bromo-2-((3-methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazol-4-yl)amino)pyrimidin-4-yl)amino)methyl)benzenesulfonamide

  • MDL number N/A
  • Cas Number : 2361659-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.ambeed.com/products/2361659-62-1.html

Safety Information

MSDS https://www.ambeed.com/products/2361659-62-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart