AMBH580642A7 Ambeed, Inc.

N1-(2-(Dimethylamino)ethyl)-2-fluorobenzene-1,4-diamine

  • MDL number MFCD11193246
  • Cas Number : 1039333-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1039333-52-2.html

Safety Information

MSDS https://www.ambeed.com/products/1039333-52-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart