AMBH580642AA Ambeed, Inc.

2-(Chloromethyl)-6-methoxy-3-(trifluoromethyl)pyridine

  • MDL number MFCD28756615
  • Cas Number : 1807153-07-6
  • Molecular Weight : 225.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(chloromethyl)-6-methoxy-3-(trifluoromethyl)pyridine
Inchi Code 1S/C8H7ClF3NO/c1-14-7-3-2-5(8(10,11)12)6(4-9)13-7/h2-3H,4H2,1H3
Inchi Key BTTFGWNPWVLNNW-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Inert atmosphere,2-8
Physical Form Solid or liquid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Product Link https://www.ambeed.com/products/1807153-07-6.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1807153-07-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart