AMBH580642AB Ambeed, Inc.

V-9302 hydrochloride

  • MDL number N/A

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.ambeed.com/products/P001057409.html

Safety Information

MSDS https://www.ambeed.com/products/P001057409.html
Availability details for
Quantity
  • Availability
  • Add to Cart