AMBH580642AC Ambeed, Inc.

O,O'-(Ethane-1,2-diyl)bis(hydroxylamine) dihydrochloride

  • MDL number MFCD15832858
  • Cas Number : 104845-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/104845-83-2.html

Safety Information

MSDS https://www.ambeed.com/products/104845-83-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart