AMBH580642AE Ambeed, Inc.

6-(6-Morpholino-1,3-dioxo-1H-benzo[de]isoquinolin-2(3H)-yl)hexanoic acid

  • MDL number MFCD02916602
  • Cas Number : 423146-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/423146-25-2.html

Safety Information

MSDS https://www.ambeed.com/products/423146-25-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart