AMBH580642AF Ambeed, Inc.

Pyridine-2,5-dicarbaldehyde

  • MDL number MFCD13175969
  • Cas Number : 6221-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/6221-01-8.html

Safety Information

MSDS https://www.ambeed.com/products/6221-01-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart