AMBH580642B3 Ambeed, Inc.

Methyl 1-acetyl-3-methyl-1H-indazole-6-carboxylate

  • MDL number MFCD32069581
  • Cas Number : 2368870-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/2368870-45-3.html

Safety Information

MSDS https://www.ambeed.com/products/2368870-45-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart