AMBH580642B5 Ambeed, Inc.

1-Benzyl-1-(1-butylpiperidin-4-yl)-3-(3,4-dichlorophenyl)urea hydrochloride

  • MDL number N/A

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/P001060723.html

Safety Information

MSDS https://www.ambeed.com/products/P001060723.html
Availability details for
Quantity
  • Availability
  • Add to Cart