AMBH580642B9 Ambeed, Inc.

tert-Butyl 2-(bromomethyl)-5-nitrobenzoate

  • MDL number MFCD32214878
  • Cas Number : 2442597-45-5
  • Molecular Weight : 316.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-(bromomethyl)-5-nitrobenzoate

Description

Purity 97%
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/2442597-45-5.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/2442597-45-5.html
pictograms GHS05,GHS07
hstatements H302-H314
Availability details for
Quantity
  • Availability
  • Add to Cart