AMBH580642BA Ambeed, Inc.

(3S,4R,5R)-3,4,5-Tris(benzyloxy)tetrahydro-2H-pyran-2-ol

  • MDL number N/A

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/P001061162.html

Safety Information

MSDS https://www.ambeed.com/products/P001061162.html
Availability details for
Quantity
  • Availability
  • Add to Cart