AMBH580642BB Ambeed, Inc.

Carbetocin acetate

  • MDL number MFCD10566502
  • Cas Number : 1936459-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/1936459-26-5.html

Safety Information

MSDS https://www.ambeed.com/products/1936459-26-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart