AMBH580642BE Ambeed, Inc.

N-(2-Chloro-4-pyridinyl)benzamide

  • MDL number MFCD23131104
  • Cas Number : 591754-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/591754-12-0.html

Safety Information

MSDS https://www.ambeed.com/products/591754-12-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart