AMBH580642BF Ambeed, Inc.

5-Nitrobenzo[d]thiazole-2-carbonitrile

  • MDL number MFCD18813185
  • Cas Number : 188672-84-6
  • Molecular Weight : 205.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/188672-84-6.html

Safety Information

MSDS https://www.ambeed.com/products/188672-84-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart