AMBH580642CA Ambeed, Inc.

4-Methyl-1-(3-nitro-5-(trifluoromethyl)phenyl)-1H-imidazole methanesulfonate

  • MDL number MFCD01926538
  • Cas Number : 917391-29-8
  • Molecular Weight : 367.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/917391-29-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/917391-29-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart