AMBH580642D5 Ambeed, Inc.

6-(Bromomethyl)benzo[d]oxazole

  • MDL number MFCD16036445
  • Cas Number : 181040-42-6
  • Molecular Weight : 212.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/181040-42-6.html

Safety Information

MSDS https://www.ambeed.com/products/181040-42-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart