AMBH580642DA Ambeed, Inc.

Copper(II) D-gluconate

  • MDL number MFCD00075297
  • Cas Number : 527-09-3
  • Molecular Weight : 453.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/527-09-3.html

Safety Information

signalword Warning
pcodes P261-P264-P270-P271-P280-P301+P312+P330-P302+P352+P312+P362+P364-P304+P340+P312-P501
MSDS https://www.ambeed.com/products/527-09-3.html
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart