AMBH580642E4 Ambeed, Inc.

Thiophene-2,5-dicarbonitrile

  • MDL number MFCD22987914
  • Cas Number : 18853-40-2
  • Molecular Weight : 134.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/18853-40-2.html

Safety Information

MSDS https://www.ambeed.com/products/18853-40-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart