AMBH580642EB Ambeed, Inc.

4-(3-Nitrophenyl)butan-2-one

  • MDL number MFCD11655148
  • Cas Number : 3506-81-8
  • Molecular Weight : 193.1992

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/3506-81-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/3506-81-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart