AMBH580642F0 Ambeed, Inc.

(R)-Methyl 2-acetamido-3-mercaptopropanoate

  • MDL number MFCD00237476
  • Cas Number : 7652-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/7652-46-2.html

Safety Information

MSDS https://www.ambeed.com/products/7652-46-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart