AMBH580642F1 Ambeed, Inc.

AM-2394

  • MDL number MFCD30533320
  • Cas Number : 1442684-77-6
  • Molecular Weight : 423.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(6'-(2-Hydroxy-2-methylpropoxy)-4-((5-methylpyridin-3-yl)oxy)-[3,3'-bipyridin]-6-yl)-3-methylurea
Purity 98
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1442684-77-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1442684-77-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart