AMBH580642F6 Ambeed, Inc.

1-(Bromomethyl)-4-ethoxy-2-fluorobenzene

  • MDL number MFCD29762292
  • Cas Number : 887338-81-0
  • Molecular Weight : 233.0775

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/887338-81-0.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/887338-81-0.html
pictograms GHS05,GHS07
hstatements H302-H314
Availability details for
Quantity
  • Availability
  • Add to Cart